Assay Details
| Gene Symbol: PSMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095920.1 | XP_020951579.1 | 421 | 85 | ||
| GenBank mRNA | AK230696.1 | - | 3 - 4 | 391 | 85 | |
| AK346574.1 | - | 3 - 4 | 373 | 85 | ||
| AK346951.1 | - | 3 - 4 | 377 | 85 | ||
| AY609897.1 | - | 3 - 4 | 391 | 85 | ||
More Information
Set Membership: |
|

PSMB2
Genomic Map