Assay Details
| Gene Symbol: TSPAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927979.5 | XP_001928014.2 | 153 | 65 | ||
| XM_005663555.3 | XP_005663612.1 | 153 | 65 | |||
| XM_021089999.1 | XP_020945658.1 | 153 | 65 | |||
| GenBank mRNA | AK230815.1 | - | 1 - 2 | 102 | 65 | |
More Information
Set Membership: |
|

TSPAN2
Genomic Map