Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091463.1 | XP_020947122.1 | 930 | 75 | ||
| XM_021091464.1 | XP_020947123.1 | 401 | 75 | |||
| GenBank mRNA | AK230901.1 | - | 4 - 5 | 342 | 75 | |
| AK231885.1 | - | 4 - 5 | 332 | 75 | ||
| AK394487.1 | - | 4 - 5 | 346 | 75 | ||
More Information
Set Membership: |
|

RBX1
Genomic Map