Assay Details
| Gene Symbol: HCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021072989.1 | XP_020928648.1 | 731 | 112 | ||
| GenBank mRNA | AK230929.1 | - | 1 | 616 | 112 | |
| AK343672.1 | - | 1 | 638 | 112 | ||
| AK397813.1 | - | 1 | 655 | 112 | ||
| AK399457.1 | - | 1 | 638 | 112 | ||
More Information
Set Membership: |
|

HCAR2
Genomic Map