Assay Details
| Gene Symbol: MMAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929194.4 | XP_001929229.1 | 187 | 57 | ||
| GenBank mRNA | AK235785.1 | - | 1 - 2 | 135 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MMAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929194.4 | XP_001929229.1 | 187 | 57 | ||
| GenBank mRNA | AK235785.1 | - | 1 - 2 | 135 | 57 | |
Set Membership: |
|