Assay Details
| Gene Symbol: MRPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925038.5 | XP_001925073.2 | 800 | 82 | ||
| GenBank mRNA | AK235954.1 | - | 6 - 7 | 770 | 82 | |
| AK392541.1 | - | 6 - 7 | 770 | 82 | ||
| AY609899.1 | - | 6 - 7 | 723 | 82 | ||
More Information
Set Membership: |
|

MRPL3
Genomic Map