Assay Details
| Gene Symbol: GRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129072.4 | XP_003129120.2 | 696 | 70 | ||
| XM_005656518.3 | XP_005656575.2 | 372 | 70 | |||
| GenBank mRNA | AK236075.1 | - | 2 - 3 | 552 | 70 | |
| AK392743.1 | - | 2 - 3 | 233 | 70 | ||
| AK396763.1 | - | 2 - 3 | 578 | 70 | ||
More Information
Set Membership: |
|

GRSF1
Genomic Map