Assay Details
| Gene Symbol: C1H6orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480211.4 | XP_003480259.1 | 83 | |||
| GenBank mRNA | AK236266.1 | - | 1 - 2 | 42 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003480211.4 transcripts.
Set Membership: |
|

C1H6orf120
Genomic Map