Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132045.3 | XP_003132093.1 | 322 | 83 | ||
| GenBank mRNA | AK236271.1 | - | 1 - 2 | 327 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132045.3 | XP_003132093.1 | 322 | 83 | ||
| GenBank mRNA | AK236271.1 | - | 1 - 2 | 327 | 83 | |
Set Membership: |
|