Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928532.6 | XP_001928567.2 | 648 | 93 | ||
| XM_005663011.3 | XP_005663068.1 | 648 | 93 | |||
| GenBank mRNA | AK236441.1 | - | 2 - 3 | 336 | 93 | |
| AK350912.1 | - | 2 - 3 | 269 | 93 | ||
More Information
Set Membership: |
|

HEY1
Genomic Map