Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069595.1 | XP_020925254.1 | 295 | 78 | ||
| XM_021069597.1 | XP_020925256.1 | 350 | 78 | |||
| XM_021069598.1 | XP_020925257.1 | 283 | 78 | |||
| XM_021069599.1 | XP_020925258.1 | 281 | 78 | |||
| GenBank mRNA | AK236587.1 | - | 2 - 3 | 269 | 78 | |
| AK389979.1 | - | 2 - 3 | 264 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021069598.1, XM_021069599.1 transcripts.
Set Membership: |
|

GYG1
Genomic Map