Assay Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357610.4 | XP_003357658.1 | 786 | 82 | ||
| XM_005656764.3 | XP_005656821.1 | 951 | 82 | |||
| XM_005656765.3 | XP_005656822.1 | 786 | 82 | |||
| XM_005656766.2 | XP_005656823.1 | 786 | 82 | |||
| XM_005656767.3 | XP_005656824.1 | 1000 | 82 | |||
| XM_021086471.1 | XP_020942130.1 | 786 | 82 | |||
| GenBank mRNA | AK236656.1 | - | 5 - 6 | 762 | 82 | |
| AK352024.1 | - | 5 - 6 | 747 | 82 | ||
| AK396801.1 | - | 5 - 6 | 962 | 82 | ||
| AK397912.1 | - | 5 - 6 | 766 | 82 | ||
More Information
Set Membership: |
|

UBFD1
Genomic Map