Assay Details
| Gene Symbol: BUD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124410.4 | XP_003124458.2 | 347 | 63 | ||
| GenBank mRNA | AK236674.1 | - | 4 - 5 | 296 | 63 | |
| AK343508.1 | - | 4 - 5 | 290 | 63 | ||
| AK346754.1 | - | 4 - 5 | 293 | 63 | ||
More Information
Set Membership: |
|

BUD23
Genomic Map