Assay Details
| Gene Symbol: PAICS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101375.1 | XP_020957034.1 | 886 | 67 | ||
| GenBank mRNA | AK236707.1 | - | 5 - 6 | 913 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PAICS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101375.1 | XP_020957034.1 | 886 | 67 | ||
| GenBank mRNA | AK236707.1 | - | 5 - 6 | 913 | 67 | |
Set Membership: |
|