Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089517.1 | XP_020945176.1 | 909 | 78 | ||
| XM_021089518.1 | XP_020945177.1 | 642 | 78 | |||
| XM_021089519.1 | XP_020945178.1 | 435 | 78 | |||
| GenBank mRNA | AK237093.1 | - | 4 - 5 | 902 | 78 | |
More Information
Set Membership: |
|

FCRLA
Genomic Map