Assay Details
| Gene Symbol: CRELD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126783.6 | XP_003126831.4 | 1044 | 104 | ||
| GenBank mRNA | AK237146.1 | - | 8 - 9 | 972 | 104 | |
| AK238194.1 | - | 9 - 10 | 1093 | 104 | ||
| AK396980.1 | - | 8 - 9 | 1049 | 104 | ||
| AK399266.1 | - | 7 - 8 | 712 | 104 | ||
More Information
Set Membership: |
|

CRELD2
Genomic Map