Assay Details
| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928070.5 | XP_001928105.1 | 517 | 68 | ||
| GenBank mRNA | AK238092.1 | - | 3 - 4 | 463 | 68 | |
| AK398419.1 | - | 3 - 4 | 503 | 68 | ||
More Information
Set Membership: |
|

SNX12
Genomic Map