Assay Details
| Gene Symbol: NDUFV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358981.5 | XP_003359029.4 | 141 | 107 | ||
| XM_021071042.1 | XP_020926701.1 | 141 | 107 | |||
| GenBank mRNA | AK238489.1 | - | 1 - 2 | 64 | 107 | |
| AK398172.1 | - | 1 - 2 | 73 | 107 | ||
More Information
Set Membership: |
|

NDUFV3
Genomic Map