Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135511.4 | XP_003135559.1 | 340 | 69 | ||
| XM_005674022.3 | XP_005674079.1 | 327 | 69 | |||
| XM_021080447.1 | XP_020936106.1 | 641 | 69 | |||
| XM_021080448.1 | XP_020936107.1 | 629 | 69 | |||
| XM_021080450.1 | XP_020936109.1 | 609 | 69 | |||
| GenBank mRNA | AK231030.1 | - | 2 - 3 | 296 | 69 | |
| AK344409.1 | - | 2 - 3 | 277 | 69 | ||
More Information
Set Membership: |
|

TMLHE
Genomic Map