Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994473.2 | XP_013849927.1 | 513 | 88 | ||
| XM_013994474.2 | XP_013849928.1 | 290 | 88 | |||
| XM_021085296.1 | XP_020940955.1 | 292 | 88 | |||
| GenBank mRNA | AK231149.1 | - | 2 - 3 | 210 | 88 | |
| AK391389.1 | - | 2 - 3 | 503 | 88 | ||
More Information
Set Membership: |
|

DNAJC24
Genomic Map