Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928466.6 | XP_001928501.1 | 1623 | 58 | ||
| XM_013981172.2 | XP_013836626.1 | 1659 | 58 | |||
| XM_021090347.1 | XP_020946006.1 | 1767 | 58 | |||
| XM_021090348.1 | XP_020946007.1 | 1659 | 58 | |||
| XM_021090349.1 | XP_020946008.1 | 1659 | 58 | |||
| XM_021090350.1 | XP_020946009.1 | 1623 | 58 | |||
| XM_021090351.1 | XP_020946010.1 | 1659 | 58 | |||
| XM_021090352.1 | XP_020946011.1 | 1659 | 58 | |||
| XM_021090353.1 | XP_020946012.1 | 926 | 58 | |||
| GenBank mRNA | AK231151.1 | - | 11 - 12 | 1466 | 58 | |
| AK399428.1 | - | 6 - 7 | 844 | 58 | ||
More Information
Set Membership: |
|

DNAI3
Genomic Map