Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928466.6 | XP_001928501.1 | 894 | 119 | ||
| XM_021090350.1 | XP_020946009.1 | 894 | 119 | |||
| GenBank mRNA | AK231151.1 | - | 6 - 7 | 737 | 119 | |
| AK399428.1 | - | 1 - 2 | 115 | 119 | ||
More Information
Set Membership: |
|

DNAI3
Genomic Map