Assay Details
| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134762.6 | XP_003134810.3 | 1092 | 88 | ||
| XM_005673228.3 | XP_005673285.1 | 1089 | 88 | |||
| XM_005673229.3 | XP_005673286.1 | 1035 | 88 | |||
| XM_021079118.1 | XP_020934777.1 | 974 | 88 | |||
| XM_021079120.1 | XP_020934779.1 | 918 | 88 | |||
| GenBank mRNA | AK231181.1 | - | 2 - 3 | 495 | 88 | |
| AK235716.1 | - | 4 - 5 | 787 | 88 | ||
| AK235909.1 | - | 4 - 5 | 1007 | 88 | ||
| AK238299.1 | - | 4 - 5 | 639 | 88 | ||
| AK238746.1 | - | 4 - 5 | 912 | 88 | ||
| AK393824.1 | - | 4 - 5 | 869 | 88 | ||
| AK395462.1 | - | 4 - 5 | 1033 | 88 | ||
| AK396265.1 | - | 4 - 5 | 1090 | 88 | ||
| AY609586.1 | - | 4 - 5 | 631 | 88 | ||
| FJ042672.1 | - | 2 - 3 | 342 | 88 | ||
More Information
Set Membership: |
|

MDFIC
Genomic Map