Assay Details
| Gene Symbol: FJX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353898.4 | XP_003353946.1 | 893 | 62 | ||
| GenBank mRNA | AK231509.1 | - | 1 | 423 | 62 | |
| AK351690.1 | - | 1 | 449 | 62 | ||
More Information
Set Membership: |
|

FJX1
Genomic Map