Assay Details
| Gene Symbol: LOC110256626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021072532.1 | XP_020928191.1 | 1641 | 72 | ||
| XM_021072533.1 | XP_020928192.1 | 4465 | 72 | |||
| XM_021072535.1 | XP_020928194.1 | 1993 | 72 | |||
| GenBank mRNA | AK231562.1 | - | 13 - 14 | 1616 | 72 | |
More Information
Set Membership: |
|

LOC110256626
Genomic Map