Assay Details
| Gene Symbol: KLK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998716.2 | XP_013854170.1 | 589 | 82 | ||
| GenBank mRNA | AK238526.1 | - | 4 - 5 | 635 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KLK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998716.2 | XP_013854170.1 | 589 | 82 | ||
| GenBank mRNA | AK238526.1 | - | 4 - 5 | 635 | 82 | |
Set Membership: |
|