Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092436.1 | XP_020948095.1 | 1335 | 71 | ||
| GenBank mRNA | AK238552.1 | - | 3 - 4 | 969 | 71 | |
More Information
Set Membership: |
|

SETBP1
Genomic Map