Assay Details
| Gene Symbol: CORO1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929345.6 | XP_001929380.1 | 766 | 63 | ||
| XM_005670752.3 | XP_005670809.2 | 731 | 63 | |||
| GenBank mRNA | AK238996.1 | - | 5 - 6 | 727 | 63 | |
| AK394457.1 | - | 5 - 6 | 728 | 63 | ||
More Information
Set Membership: |
|

CORO1C
Genomic Map