Assay Details
| Gene Symbol: SERTAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125089.3 | XP_003125137.2 | 3631 | 78 | ||
| XM_005662514.3 | XP_005662571.2 | 3970 | 78 | |||
| XM_013996103.2 | XP_013851557.2 | 3670 | 78 | |||
| GenBank mRNA | AK239017.1 | - | 1 | 327 | 78 | |
More Information
Set Membership: |
|

SERTAD2
Genomic Map