Assay Details
| Gene Symbol: SMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079653.1 | XP_020935312.1 | 294 | 64 | ||
| GenBank mRNA | AK239096.1 | - | 1 - 2 | 162 | 64 | |
| AK397445.1 | - | 1 - 2 | 183 | 64 | ||
More Information
Set Membership: |
|

SMS
Genomic Map