Assay Details
| Gene Symbol: PTPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096020.1 | XP_020951679.1 | 766 | 67 | ||
| XM_021096021.1 | XP_020951680.1 | 379 | 67 | |||
| XM_021096022.1 | XP_020951681.1 | 769 | 67 | |||
| XM_021096023.1 | XP_020951682.1 | 767 | 67 | |||
| GenBank mRNA | AK239437.1 | - | 3 - 4 | 359 | 67 | |
| AY610205.1 | - | 3 - 4 | 346 | 67 | ||
| EU753191.1 | - | 3 - 4 | 246 | 67 | ||
More Information
Set Membership: |
|

PTPN2
Genomic Map