Assay Details
| Gene Symbol: PITPNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657014.3 | XP_005657071.1 | 317 | 65 | ||
| XM_021067561.1 | XP_020923220.1 | 317 | 65 | |||
| XM_021067562.1 | XP_020923221.1 | 317 | 65 | |||
| GenBank mRNA | AK239695.1 | - | 2 - 3 | 269 | 65 | |
| AK389579.1 | - | 2 - 3 | 295 | 65 | ||
More Information
Set Membership: |
|

PITPNA
Genomic Map