Assay Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013988588.2 | XP_013844042.2 | 862 | 61 | ||
GenBank mRNA | AK239732.1 | - | 2 - 3 | 1129 | 61 | |
AK239929.1 | - | 1 - 2 | 73 | 61 | ||
AK396268.1 | - | 1 - 2 | 300 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
