Assay Details
| Gene Symbol: RNPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481098.4 | XP_003481146.1 | 465 | 60 | ||
| XM_003481099.4 | XP_003481147.1 | 1046 | 60 | |||
| XM_013987838.2 | XP_013843292.1 | 1241 | 60 | |||
| XM_013987839.2 | XP_013843293.1 | 308 | 60 | |||
| GenBank mRNA | AK239758.1 | - | 3 - 4 | 765 | 60 | |
| AK395890.1 | - | 3 - 4 | 405 | 60 | ||
More Information
Set Membership: |
|

RNPS1
Genomic Map