Assay Details
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926580.6 | XP_001926615.2 | 1736 | 67 | ||
| XM_005671562.3 | XP_005671619.2 | 1754 | 67 | |||
| XM_021072595.1 | XP_020928254.1 | 1710 | 67 | |||
| XM_021072596.1 | XP_020928255.1 | 2077 | 67 | |||
| XM_021072598.1 | XP_020928257.1 | 1473 | 67 | |||
| XM_021072599.1 | XP_020928258.1 | 1598 | 67 | |||
| GenBank mRNA | AK239979.1 | - | 1 - 2 | 49 | 67 | |
| AK393530.1 | - | 2 - 3 | 343 | 67 | ||
More Information
Set Membership: |
|

DHX32
Genomic Map