Assay Details
| Gene Symbol: PRMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355999.4 | XP_003356047.1 | 152 | 58 | ||
| GenBank mRNA | AK240192.1 | - | 1 - 2 | 84 | 58 | |
| AK352215.1 | - | 1 - 2 | 84 | 58 | ||
| KU504627.1 | - | 1 - 2 | 61 | 58 | ||
More Information
Set Membership: |
|

PRMT1
Genomic Map