Assay Details
| Gene Symbol: CENPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077378.1 | XP_020933037.1 | 1977 | 94 | ||
| GenBank mRNA | AK240234.1 | - | 1 | 317 | 94 | |
| AK394990.1 | - | 1 | 578 | 94 | ||
More Information
Set Membership: |
|

CENPB
Genomic Map