Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005669186.3 | XP_005669243.1 | 1307 | 89 | ||
| XM_005669188.3 | XP_005669245.1 | 1229 | 89 | |||
| XM_021067802.1 | XP_020923461.1 | 1212 | 89 | |||
| XM_021067803.1 | XP_020923462.1 | 1229 | 89 | |||
| XM_021067804.1 | XP_020923463.1 | 1222 | 89 | |||
| XM_021067805.1 | XP_020923464.1 | 1293 | 89 | |||
| GenBank mRNA | AK240317.1 | - | 3 - 4 | 1124 | 89 | |
| AK345492.1 | - | 4 - 5 | 1224 | 89 | ||
| AK347798.1 | - | 4 - 5 | 1227 | 89 | ||
| AK394877.1 | - | 4 - 5 | 1266 | 89 | ||
More Information
Set Membership: |
|

SLC16A11
Genomic Map