Assay Details
| Gene Symbol: CDC42EP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094975.1 | XP_020950634.1 | 537 | 93 | ||
| GenBank mRNA | AK240320.1 | - | 1 - 2 | 219 | 93 | |
| AK240502.1 | - | 1 - 2 | 217 | 93 | ||
| AK398685.1 | - | 1 - 2 | 232 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021094975.1 transcripts.
Set Membership: |
|

CDC42EP5
Genomic Map