Assay Details
| Gene Symbol: RBAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653058.3 | XP_005653115.2 | 330 | 88 | ||
| XM_021086052.1 | XP_020941711.1 | 581 | 88 | |||
| GenBank mRNA | AK240392.1 | - | 1 - 2 | 135 | 88 | |
More Information
Set Membership: |
|

RBAK
Genomic Map