Assay Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068740.1 | XP_020924399.1 | 5345 | 111 | ||
| XM_021068741.1 | XP_020924400.1 | 4547 | 111 | |||
| XM_021068742.1 | XP_020924401.1 | 4410 | 111 | |||
| XM_021068743.1 | XP_020924402.1 | 5340 | 111 | |||
| XM_021068744.1 | XP_020924403.1 | 5288 | 111 | |||
| XM_021068746.1 | XP_020924405.1 | 4736 | 111 | |||
| XM_021068747.1 | XP_020924406.1 | 4672 | 111 | |||
| XM_021068748.1 | XP_020924407.1 | 4702 | 111 | |||
| XM_021068749.1 | XP_020924408.1 | 4877 | 111 | |||
| XM_021068750.1 | XP_020924409.1 | 5093 | 111 | |||
| XM_021068751.1 | XP_020924410.1 | 4624 | 111 | |||
| XM_021068752.1 | XP_020924411.1 | 4837 | 111 | |||
| XM_021068753.1 | XP_020924412.1 | 4479 | 111 | |||
| XM_021068754.1 | XP_020924413.1 | 4835 | 111 | |||
| XM_021068755.1 | XP_020924414.1 | 4497 | 111 | |||
| XM_021068757.1 | XP_020924416.1 | 4665 | 111 | |||
| XM_021068758.1 | XP_020924417.1 | 4577 | 111 | |||
| XM_021068759.1 | XP_020924418.1 | 4424 | 111 | |||
| XM_021068760.1 | XP_020924419.1 | 4597 | 111 | |||
| GenBank mRNA | AK240563.1 | - | 1 | 122 | 111 | |
| AK394951.1 | - | 1 | 380 | 111 | ||
More Information
Set Membership: |
|

DAG1
Genomic Map