Assay Details
| Gene Symbol: NEURL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926195.5 | XP_001926230.2 | 1820 | 136 | ||
| GenBank mRNA | AK343471.1 | - | 1 - 2 | 1475 | 136 | |
| AY609743.1 | - | 1 - 2 | 812 | 136 | ||
More Information
Set Membership: |
|

NEURL2
Genomic Map