Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133332.5 | XP_003133380.3 | 1451 | 81 | ||
| XM_021074832.1 | XP_020930491.1 | 1470 | 81 | |||
| GenBank mRNA | AK343600.1 | - | 1 - 2 | 381 | 81 | |
More Information
Set Membership: |
|

CDKN2AIP
Genomic Map