Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362134.4 | XP_003362182.1 | 465 | 82 | ||
| GenBank mRNA | AK343742.1 | - | 5 - 6 | 429 | 82 | |
More Information
Set Membership: |
|

BORCS8
Genomic Map