Assay Details
| Gene Symbol: PIGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355104.4 | XP_003355152.1 | 1223 | 105 | ||
| GenBank mRNA | AK344279.1 | - | 1 | 1116 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PIGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355104.4 | XP_003355152.1 | 1223 | 105 | ||
| GenBank mRNA | AK344279.1 | - | 1 | 1116 | 105 | |
Set Membership: |
|