Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132703.3 | XP_003132751.1 | 768 | 80 | ||
| XM_013981108.2 | XP_013836562.1 | 548 | 80 | |||
| GenBank mRNA | AK344332.1 | - | 3 - 4 | 709 | 80 | |
More Information
Set Membership: |
|

TMEM45A
Genomic Map