Assay Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128956.4 | XP_003129004.1 | 458 | 98 | ||
| XM_021100563.1 | XP_020956222.1 | 458 | 98 | |||
| XM_021100564.1 | XP_020956223.1 | 458 | 98 | |||
| GenBank mRNA | AK344384.1 | - | 3 - 4 | 387 | 98 | |
More Information
Set Membership: |
|

COMMD8
Genomic Map