Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021072526.1 | XP_020928185.1 | 696 | 59 | ||
| XM_021072527.1 | XP_020928186.1 | 608 | 59 | |||
| XM_021072528.1 | XP_020928187.1 | 696 | 59 | |||
| XM_021072529.1 | XP_020928188.1 | 696 | 59 | |||
| XM_021072530.1 | XP_020928189.1 | 696 | 59 | |||
| XM_021072531.1 | XP_020928190.1 | 342 | 59 | |||
| GenBank mRNA | AK344408.1 | - | 3 - 4 | 342 | 59 | |
More Information
Set Membership: |
|

CCDC63
Genomic Map