Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996877.2 | XP_013852331.2 | 399 | 57 | ||
| GenBank mRNA | AK344624.1 | - | 1 - 2 | 147 | 57 | |
More Information
Set Membership: |
|

SLAMF9
Genomic Map