Assay Details
| Gene Symbol: TSPYL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089070.1 | XP_020944729.1 | 2894 | 108 | ||
| XM_021089071.1 | XP_020944730.1 | 2867 | 108 | |||
| XM_021089072.1 | XP_020944731.1 | 2816 | 108 | |||
| GenBank mRNA | AK344733.1 | - | 1 | 123 | 108 | |
More Information
Set Membership: |
|

TSPYL5
Genomic Map